Methyl 1,2,4-triazole-3-carboxylate, 98%
Methyl 1,2,4-triazole-3-carboxylate, 98%
Methyl 1,2,4-triazole-3-carboxylate, 98%
Thermo Scientific Chemicals

Methyl 1,2,4-triazole-3-carboxylate, 98%

CAS: 4928-88-5 | C4H5N3O2 | 127.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 415000250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS4928-88-5
IUPAC Namemethyl 1H-1,2,4-triazole-5-carboxylate
Molecular FormulaC4H5N3O2
InChI KeyQMPFMODFBNEYJH-UHFFFAOYSA-N
SMILESCOC(=O)C1=NC=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Melting point185°C to 193°C
HPLC>=97.5 %
Appearance (Form)Powder and/or flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only