Dimethyl trisulfide, 98+%
Dimethyl trisulfide, 98+%
Dimethyl trisulfide, 98+%
Dimethyl trisulfide, 98+%
Thermo Scientific Chemicals

Dimethyl trisulfide, 98+%

CAS: 3658-80-8 | C2H6S3 | 126.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 415030250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3658-80-8
IUPAC Namedimethyltrisulfane
Molecular FormulaC2H6S3
InChI KeyYWHLKYXPLRWGSE-UHFFFAOYSA-N
SMILESCSSSC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Refractive index1.5995 to 1.6025 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only