Methyl Violet 2B, pure, certified
Methyl Violet 2B, pure, certified
Methyl Violet 2B, pure, certified
Thermo Scientific Chemicals

Methyl Violet 2B, pure, certified

CAS: 8004-87-3 | C24H28ClN3
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Glass bottle
Catalog number 415075000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS8004-87-3
IUPAC Namebis[4-(dimethylamino)phenyl][4-(methylamino)phenyl]methylium
Molecular FormulaC24H28N3
InChI KeyHJPVVBCICYOONZ-UHFFFAOYSA-N
SMILESCNC1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Dye content>=75 %
UV Lambda max582 to 588 nm
Absorption Ratio P-15/P+151.06 to 1.19
BSC CertificationPasses
Appearance (Color)Green
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only