Mineral oil, pure
Thermo Scientific Chemicals

Mineral oil, pure

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
6 mL
1 L
2.5 L
10 L
Catalog number 415080025
Price (USD)
113.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
113.00
Each
Chemical Identifiers
CAS8042-47-5
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4730 to 1.4800 (20°C, 589 nm)
Density(15°C) 0.860 to 0.880 g/ml
Viscosity63 to 80 mm²/s (40°C)
Appearance (Form)Oily liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only