Ensure optimal chromatography performance and cost savings with Hypersil PREP HS Phenyl bulk chromatography media.
Have Questions?
Change view
Catalog Number
Quantity
41510-00500B
500 g
41510-00050B
50 g
41510-00100B
100 g
41510-01000B
1000 g
41510-05000B
5000 g
41510-10000B
10000 g
6 Options
Catalog number 41510-00500B
Price (USD)
-
Quantity:
500 g
Ensure optimal chromatography performance with our Thermo Scientific™ Hypersil™ PREP HS Phenyl bulk chromatography media. Purchase bulk quantities of moderately hydrophobic, high surface area, phenyl-bonded media to repack preparative HPLC columns yourself without paying for new hardware.
Repack Columns Yourself with Bulk Media
Unique Selectivity
Enhanced selectivity for aromatic and moderately polar analytes.
Selectivity complementary to that provided by alkyl chain phases.
Moderate hydrophobicity compared to C18.
Phenyl (USP L11) selectivity - high surface area particle with 100Å pore size.
pH stable from 2 to 8.
Maximum temperature of 60°C.
Customer Specific Orders
Please contact your local sales representative for Customer Specific Orders.