Naphthenic acids, pract.
Naphthenic acids, pract.
Naphthenic acids, pract.
Thermo Scientific Chemicals

Naphthenic acids, pract.

CAS: 1338-24-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 415280025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS1338-24-5
IUPAC Name3-(3-ethylcyclopentyl)propanoic acid
Molecular FormulaC10H18O2
InChI KeyWVRFSLWCFASCIS-UHFFFAOYNA-N
SMILESCCC1CCC(CCC(O)=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Density(20°C) 910 to 940 g/l
Acid value230 to 260 mg KOH/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only