Neutral Red, pure, certified
Neutral Red, pure, certified
Neutral Red, pure, certified
Thermo Scientific Chemicals

Neutral Red, pure, certified

CAS: 553-24-2 | C15H17ClN4 | 288.779 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 415490250
Price (USD)
124.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
124.00
Each
Chemical Identifiers
CAS553-24-2
IUPAC Namehydrogen N2,N2,7-trimethylphenazine-2,8-diamine chloride
Molecular FormulaC15H17ClN4
InChI KeyPGSADBUBUOPOJS-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Dye content>=50 %
UV Lambda max539 to 544 nm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only