Nickel(II) acetate tetrahydrate, 99%, extra pure
Nickel(II) acetate tetrahydrate, 99%, extra pure
Nickel(II) acetate tetrahydrate, 99%, extra pure
Thermo Scientific Chemicals

Nickel(II) acetate tetrahydrate, 99%, extra pure

CAS: 6018-89-9 | C4H14NiO8 | 248.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 415540050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS6018-89-9
IUPAC Namenickel(2+) diacetate tetrahydrate
Molecular FormulaC4H14NiO8
InChI KeyOINIXPNQKAZCRL-UHFFFAOYSA-L
SMILESO.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Assay>=98.5 %
Cobalt (Co)=<0.2 %
Iron (Fe)=<20 ppm
Appearance (Color)Green to blue
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only