Nickel(II) oxide, 97%, green, reagent, powder
Nickel(II) oxide, 97%, green, reagent, powder
Nickel(II) oxide, 97%, green, reagent, powder
Thermo Scientific Chemicals

Nickel(II) oxide, 97%, green, reagent, powder

CAS: 1313-99-1 | NiO | 74.69 g/mol
Have Questions?
Catalog NumberQuantityPackaging
415581000100 gGlass bottle
Catalog number 415581000
Price (USD)
79.90
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
79.90
Each
Chemical Identifiers
CAS1313-99-1
IUPAC Nameoxonickel
Molecular FormulaNiO
InChI KeyGNRSAWUEBMWBQH-UHFFFAOYSA-N
SMILESO=[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Copper (Cu)=<0.0005 %
Magnesium (Mg)=<0.01 %
Manganese (Mn)=<0.001 %
Appearance (Color)Green
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only