1-Octanesulfonic Acid, Sodium Salt Anhydrous, HPLC Grade, Thermo Scientific Chemicals
1-Octanesulfonic Acid, Sodium Salt Anhydrous, HPLC Grade, Thermo Scientific Chemicals
1-Octanesulfonic Acid, Sodium Salt Anhydrous, HPLC Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Octanesulfonic Acid, Sodium Salt Anhydrous, HPLC Grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 416361000
Price (USD)
960.65
Online Exclusive
1,067.00
Save 106.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
960.65
Online Exclusive
1,067.00
Save 106.35 (10%)
Each
Chemical Identifiers
CAS5324-84-5
IUPAC Namesodium octane-1-sulfonate
Molecular FormulaC8H17NaO3S
InChI KeyHRQDCDQDOPSGBR-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 210 nm A: =<1.00
UVat 220 nm A: =<0.10
UVat 254 nm A: =<0.04
UVat 280 nm A: =<0.04
Appearance (Form)Crystalline solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only