Olive oil, pure, virgin
Thermo Scientific Chemicals

Olive oil, pure, virgin

CAS: 8001-25-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 416545000
Price (USD)
71.10
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
71.10
Each
Chemical Identifiers
CAS8001-25-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to yellow-green
Appearance (Form)Oily liquid
Water=<0.1 %
Composition0.5 to 5.0 % (Stearic acid)
Composition2.5 to 21.0 % (Linoleic acid)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only