Orange G, pure, certified
Orange G, pure, certified
Orange G, pure, certified
Thermo Scientific Chemicals

Orange G, pure, certified

CAS: 1936-15-8 | C16H10N2Na2O7S2 | 452.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 416551000
Price (USD)
159.65
Special offer
Online exclusive
Ends: 28-Dec-2026
188.00
Save 28.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
159.65
Special offer
Online exclusive
Ends: 28-Dec-2026
188.00
Save 28.35
Each
Chemical Identifiers
CAS1936-15-8
IUPAC Namedisodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
Molecular FormulaC16H10N2Na2O7S2
InChI KeyAEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-red to red
Infrared spectrumConforms
UV Lambda max476 to 481 nm
Dye content>=80 %
Absorption Ratio P-15/P+150.88 to 0.95
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only