Palmitic acid, sodium salt, 98%
Palmitic acid, sodium salt, 98%
Palmitic acid, sodium salt, 98%
Thermo Scientific Chemicals

Palmitic acid, sodium salt, 98%

CAS: 408-35-5 | C16H31NaO2 | 278.4 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 416700050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS408-35-5
IUPAC Namesodium hexadecanoate
Molecular FormulaC16H31NaO2
InChI KeyGGXKEBACDBNFAF-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCCCCC([O-])=O
さらに表示
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Loss on drying=<2 % (110°C, 4 hrs)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only