Papain, for Biochemistry - FAQs
View additional product information for Papain, for Biochemistry - FAQs (416760100, 416761000, 416765000)
1 product FAQs found
What does the unit TU/mg mean for enzyme activity versus the commonly used unit U/mg?
Papain activity is measured in terms of units of tyrosine release per mg (TU/mg), and this unit can be used interchangeably with the more general units per mg (U/mg).