Pectin, pract.
Thermo Scientific Chemicals

Pectin, pract.

CAS: 9000-69-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 416862500
Price (USD)
128.00
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
128.00
Each
Chemical Identifiers
CAS9000-69-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige to golden
Appearance (Form)Powder
Viscosity30.5 to 36.5
Assay67 to 71 % (degree of esterification)
Loss on drying=<12 % (105°C, 2 hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only