Petrolatum, white, pure
Thermo Scientific Chemicals

Petrolatum, white, pure

CAS: 8009-03-8 | C15H15N | 209.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 417090100
Price (USD)
-
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS8009-03-8
SMILESCC1=NC2=CC=C3C=CC=CC3=C2C1(C)C
Synonympetroleum jelly
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Paste
Infrared spectrumConforms
Solidification point50°C to 56°C
Viscosity5 to 8 mm²/s (at 100°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only