Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Thermo Scientific Chemicals

Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000

CAS: 26837-42-3 | (C2H3KO4S)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 418141000
Price (USD)
2,618.65
Special offer
Online exclusive
Ends: 28-Dec-2026
3,081.00
Save 462.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
2,618.65
Special offer
Online exclusive
Ends: 28-Dec-2026
3,081.00
Save 462.35
Each
Chemical Identifiers
CAS26837-42-3
Molecular Formula(C2H3KO4S)n
SMILES[K]OS(=O)(=O)OC(-*)C-*
MDL NumberMFCD00081923
SpecificationsSpecification SheetSpecification Sheet
Potassium (K)>=21.2 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only