2-Propene-1-sulfonic acid sodium salt, 95%, pract.
2-Propene-1-sulfonic acid sodium salt, 95%, pract.
2-Propene-1-sulfonic acid sodium salt, 95%, pract.
Thermo Scientific Chemicals

2-Propene-1-sulfonic acid sodium salt, 95%, pract.

CAS: 2495-39-8 | C3H5NaO3S | 144.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
41837025025 gGlass bottle
Catalog number 418370250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2495-39-8
IUPAC Namesodium prop-2-ene-1-sulfonate
Molecular FormulaC3H5NaO3S
InChI KeyDIKJULDDNQFCJG-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration after Ion exchange>=94 %
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only