1-Pyrenebutyric acid, 97+%
1-Pyrenebutyric acid, 97+%
1-Pyrenebutyric acid, 97+%
Thermo Scientific Chemicals

1-Pyrenebutyric acid, 97+%

CAS: 3443-45-6 | C20H16O2 | 288.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 418510010
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.70
Save 11.05
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 28-Dec-2026
73.70
Save 11.05
Each
Chemical Identifiers
CAS3443-45-6
SpecificationsSpecification SheetSpecification Sheet
Melting point185°C to 190°C
Appearance (Color)Light yellow to beige
Infrared spectrumConforms
HPLC>=97.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only