Pyronin Y, pure, certified
Pyronin Y, pure, certified
Pyronin Y, pure, certified
Thermo Scientific Chemicals

Pyronin Y, pure, certified

CAS: 92-32-0 | C17H19ClN2O | 302.802 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 418630010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS92-32-0
IUPAC Name3,6-bis(dimethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC17H19ClN2O
InChI KeyINCIMLINXXICKS-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC2=[O+]C3=CC(=CC=C3C=C2C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green
Infrared spectrumConforms
Absorption Ratio P-15/P+151.23 to 1.7
UV Lambda max546 to 551 nm
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only