13-cis-Retinoic acid, 99%
13-cis-Retinoic acid, 99%
13-cis-Retinoic acid, 99%
Thermo Scientific Chemicals

13-cis-Retinoic acid, 99%

CAS: 4759-48-2 | C20H28O2 | 300.44 g/mol
Have Questions?
Catalog NumberQuantityPackaging
418975000500 mgGlass bottle
Catalog number 418975000
Price (USD)
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4759-48-2
IUPAC Name(2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
Molecular FormulaC20H28O2
InChI KeySHGAZHPCJJPHSC-XFYACQKRSA-N
SMILESCC(C=CC1=C(C)CCCC1(C)C)=C/C=C/C(/C)=CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only