Rhodamine 110, 98+%, pure, laser grade
Rhodamine 110, 98+%, pure, laser grade
Rhodamine 110, 98+%, pure, laser grade
Thermo Scientific Chemicals

Rhodamine 110, 98+%, pure, laser grade

CAS: 13558-31-1 | C20H15ClN2O3 | 366.801 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Catalog number 419075000
Price (USD)
178.00
Each
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
178.00
Each
Chemical Identifiers
CAS13558-31-1
IUPAC Name3,6-diamino-9-(2-carboxyphenyl)-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC20H15ClN2O3
InChI KeyMYIOYATURDILJN-UHFFFAOYSA-N
SMILES[Cl-].NC1=CC2=[O+]C3=CC(N)=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to red to red-brown
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
HPLC>=98.0 %
UV Lambda max498 to 504 nm (c=10 mg/l in Methanol)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only