Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh
Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh
Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh
Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh
Thermo Scientific Chemicals

Rhodium(III) chloride, anhydrous, 48.6-49.2% Rh

CAS: 10049-07-7 | Cl3Rh | 209.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Catalog number 419105000
Price (TWD)
-
Request A Quote
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10049-07-7
IUPAC Namerhodium(3+) trichloride
Molecular FormulaCl3Rh
InChI KeySONJTKJMTWTJCT-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Rh+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red, grey, red-black or black
Appearance (Form)Crystalline powder, crystals or granules
Assay48.6 to 49.2 % (Rh)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only