Selenium, 99+%, powder
Selenium, 99+%, powder
Selenium, 99+%, powder
Thermo Scientific Chemicals

Selenium, 99+%, powder

CAS: 7782-49-2 | Se | 78.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 419271000
Price (EUR)
105,65
Online exclusive
117,00
Save 11,35 (10%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
105,65
Online exclusive
117,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS7782-49-2
IUPAC Nameselenium
Molecular FormulaSe
InChI KeyBUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES[Se]
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<500 ppm
Nickel (Ni)=<100 ppm
Lead (Pb)=<500 ppm
Appearance (Color)Dark grey to black
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only