Sodium iodate, 99%, extra pure, reagent, powder
Sodium iodate, 99%, extra pure, reagent, powder
Sodium iodate, 99%, extra pure, reagent, powder
Thermo Scientific Chemicals

Sodium iodate, 99%, extra pure, reagent, powder

CAS: 7681-55-2 | INaO3 | 197.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
Packaging:
Glass bottle
Catalog number 419560050
Price (USD)
23.65
Special offer
Online exclusive
Ends: 28-Dec-2026
28.00
Save 4.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
23.65
Special offer
Online exclusive
Ends: 28-Dec-2026
28.00
Save 4.35
Each
Chemical Identifiers
CAS7681-55-2
IUPAC Namesodium iodate
Molecular FormulaINaO3
InChI KeyWTCBONOLBHEDIL-UHFFFAOYSA-M
SMILES[Na+].[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<5 ppm
pH5.0 to 8.0 (5 % at 20°C)
Insoluble matter=<0.005 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only