Sodium metabisulfite, 97+%, ACS reagent
Sodium metabisulfite, 97+%, ACS reagent
Sodium metabisulfite, 97+%, ACS reagent
Thermo Scientific Chemicals

Sodium metabisulfite, 97+%, ACS reagent

CAS: 7681-57-4 | Na2O5S2 | 190.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 419580010
Price (USD)
113.65
Special offer
Online exclusive
Ends: 28-Dec-2026
134.00
Save 20.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 28-Dec-2026
134.00
Save 20.35
Each
Chemical Identifiers
CAS7681-57-4
IUPAC Namedisodium (sulfinatooxy)sulfinate
Molecular FormulaNa2O5S2
InChI KeyLDTLADDKFLAYJA-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)OS([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Iodimetric>=97.0 %
Heavy metals (as Pb)=<10 ppm
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only