DMEM, high glucose, pyruvate, 500 mL - FAQs
DMEM, high glucose, pyruvate, 500 mL - FAQs
View additional product information for DMEM, high glucose, pyruvate - FAQs (41966029)
2 product FAQs found
What is the difference between DMEM, high glucose, pyruvate Cat. No. 41966029 and Cat. No. 11995065?
Cat. No. 41966029 is manufactured in the Paisley, Scotland facility, while Cat. No. 11995065 is dual manufactured in Grand Island, New York and Paisley, Scotland. Although the formulation is said to be equivalent for molarities, the components may differ (e.g., it could be that one used L-Cystine 2HCl the other L-Cystine). This should not affect performance. Both facilities are FDA-registered and ISO 13485 certified.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?
Manganese is not present in the formulation of our catalog DMEM media products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.