Sulforhodamine 101 (free acid), 99%, pure, laser grade
Sulforhodamine 101 (free acid), 99%, pure, laser grade
Sulforhodamine 101 (free acid), 99%, pure, laser grade
Thermo Scientific Chemicals

Sulforhodamine 101 (free acid), 99%, pure, laser grade

CAS: 60311-02-6 | C31H29ClN2O6S2 | 625.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
2.5 g
Catalog number 419940025
Price (USD)
1,149.65
Online Exclusive
1,277.00
Save 127.35 (10%)
Each
Quantity:
2.5 g
Request bulk or custom format
Price (USD)
1,149.65
Online Exclusive
1,277.00
Save 127.35 (10%)
Each
Chemical Identifiers
CAS60311-02-6
IUPAC Name16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3λ⁴-oxa-9,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium
Molecular FormulaC31H29ClN2O6S2
InChI KeyMPLHNVLQVRSVEE-UHFFFAOYSA-N
SMILES[O-]S(=O)(=O)C1=CC(=CC=C1C1=C2C=C3CCCN4CCCC(C2=[O+]C2=C5CCCN6CCCC(C=C12)=C56)=C34)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=98.5 %
Molar absorption coefficient>=105000 l/(mol.cm) (at Lambda max)
Suitability(for use as a laser dye) Passes test
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only