Sulfuric acid, extra pure, fuming, 20-30% free SO3
Sulfuric acid, extra pure, fuming, 20-30% free SO3
Sulfuric acid, extra pure, fuming, 20-30% free SO3
Thermo Scientific Chemicals

Sulfuric acid, extra pure, fuming, 20-30% free SO3

CAS: 8014-95-7 | H2O7S2 | 178.13 g/mol
数量:
25 g
500 g
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 419970250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS8014-95-7
IUPAC Namesulfonylideneoxidane; sulfuric acid
Molecular FormulaH2O7S2
InChI KeyHIFJUMGIHIZEPX-UHFFFAOYSA-N
SMILESO=S(=O)=O.OS(O)(=O)=O
さらに表示
Appearance (Form)Liquid
Ammonium (NH4)=<30 ppm
Assay20 to 30 % (free SO3)
Iron (Fe)=<50 ppm
Appearance (Color)Clear colorless to light brown
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only