CD8a Monoclonal Antibody (53-6.7), eFluor™ 615, eBioscience™ - FAQs
CD8a Monoclonal Antibody (53-6.7), eFluor™ 615, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (53-6.7), eFluor™ 615, eBioscience™ - FAQs (42-0081-82)