CD11c Monoclonal Antibody (N418), eFluor™ 615, eBioscience™ - FAQs
CD11c Monoclonal Antibody (N418), eFluor™ 615, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (N418), eFluor™ 615, eBioscience™ - FAQs (42-0114-82)