CD15 Monoclonal Antibody (HI98), eFluor™ 615, eBioscience™ - FAQs
CD15 Monoclonal Antibody (HI98), eFluor™ 615, eBioscience™ - FAQs
View additional product information for CD15 Monoclonal Antibody (HI98), eFluor™ 615, eBioscience™ - FAQs (42-0159-82, 42-0159-80)