CD20 Monoclonal Antibody (L26), eFluor™ 615, eBioscience™ - FAQs
CD20 Monoclonal Antibody (L26), eFluor™ 615, eBioscience™ - FAQs
View additional product information for CD20 Monoclonal Antibody (L26), eFluor™ 615, eBioscience™ - FAQs (42-0202-80)