Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 615, eBioscience™ - FAQs
Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 615, eBioscience™ - FAQs
View additional product information for Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 615, eBioscience™ - FAQs (42-4301-80)