eBioscience™ Streptavidin eFluor™ 615 Conjugate, 100 μg - FAQs
eBioscience™ Streptavidin eFluor™ 615 Conjugate, 100 μg - FAQs
View additional product information for eBioscience™ Streptavidin eFluor™ 615 Conjugate - FAQs (42-4317-80, 42-4317-82)