Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 615, eBioscience™ - FAQs
Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 615, eBioscience™ - FAQs
View additional product information for Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 615, eBioscience™ - FAQs (42-4321-80, 42-4321-82)