Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 615, eBioscience™ - FAQs
Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 615, eBioscience™ - FAQs
View additional product information for Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 615, eBioscience™ - FAQs (42-4724-80)