Mouse anti-Rat IgG1 Secondary Antibody, eFluor™ 615, eBioscience™ - FAQs
Mouse anti-Rat IgG1 Secondary Antibody, eFluor™ 615, eBioscience™ - FAQs
View additional product information for Mouse anti-Rat IgG1 Secondary Antibody, eFluor™ 615, eBioscience™ - FAQs (42-4812-82)