Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 615, eBioscience™ - FAQs
Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 615, eBioscience™ - FAQs
View additional product information for Mouse anti-Rat IgG2a Secondary Antibody, eFluor™ 615, eBioscience™ - FAQs (42-4817-80, 42-4817-82)