Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 615, eBioscience™ - FAQs
Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 615, eBioscience™ - FAQs
View additional product information for Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 615, eBioscience™ - FAQs (42-4888-80)