Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 615, eBioscience™ - FAQs
Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 615, eBioscience™ - FAQs
View additional product information for Pan Cytokeratin Monoclonal Antibody (AE1/AE3), eFluor™ 615, eBioscience™ - FAQs (42-9003-80)