Cytokeratin 7 Monoclonal Antibody (LP5K), eFluor™ 615, eBioscience™ - FAQs
Cytokeratin 7 Monoclonal Antibody (LP5K), eFluor™ 615, eBioscience™ - FAQs
View additional product information for Cytokeratin 7 Monoclonal Antibody (LP5K), eFluor™ 615, eBioscience™ - FAQs (42-9005-82, 42-9005-80)