PCNA Monoclonal Antibody (PC10 (3F81)), eFluor™ 615, eBioscience™ - FAQs
PCNA Monoclonal Antibody (PC10 (3F81)), eFluor™ 615, eBioscience™ - FAQs
View additional product information for PCNA Monoclonal Antibody (PC10 (3F81)), eFluor™ 615, eBioscience™ - FAQs (42-9910-82)