Magnesium sulfate heptahydrate, 99-100.5%, FCC, USP for parenteral use, GMP, Macron Fine Chemicals™
Avantor™

Magnesium sulfate heptahydrate, 99-100.5%, FCC, USP for parenteral use, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets U.S.P Requirements.Read more
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
12 kg
Catalog number 4200-07
Price (USD)
-
Quantity:
2.5 kg
This product is GMP Manufactured and meets U.S.P Requirements.
Specifications
CAS10034-99-8
Chemical Name or MaterialMagnesium sulfate heptahydrate
ColorColorless
Melting Point1124°C
Molecular FormulaMgSO4·7H2O
PackagingPoly bottle
SynonymEpsom salts
GradeUSP
Molecular Weight (g/mol)246.48
Percent Purity99-100.5%
Physical FormCrystalline
Quantity2.5 kg
Unit SizeEach