Tellurium, 99+%, (trace metal basis), powder
Tellurium, 99+%, (trace metal basis), powder
Tellurium, 99+%, (trace metal basis), powder
Thermo Scientific Chemicals

Tellurium, 99+%, (trace metal basis), powder

CAS: 13494-80-9 | Te | 127.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 420021000
Price (USD)
122.65
Online Exclusive
136.00
Save 13.35 (10%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
122.65
Online Exclusive
136.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS13494-80-9
IUPAC Nametellurium
Molecular FormulaTe
InChI KeyPORWMNRCUJJQNO-UHFFFAOYSA-N
SMILES[Te]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to black
Appearance (Form)Powder
Total trace metal impurities=<1.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only