Tetrachlorocyclopropene, 98%
Tetrachlorocyclopropene, 98%
Tetrachlorocyclopropene, 98%
Thermo Scientific Chemicals

Tetrachlorocyclopropene, 98%

CAS: 6262-42-6 | C3Cl4 | 177.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 420190250
Price (USD)
589.65
Online Exclusive
655.00
Save 65.35 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
589.65
Online Exclusive
655.00
Save 65.35 (10%)
Each
Chemical Identifiers
CAS6262-42-6
IUPAC Nametetrachlorocycloprop-1-ene
Molecular FormulaC3Cl4
InChI KeyBLZOHTXDDOAASQ-UHFFFAOYSA-N
SMILESClC1=C(Cl)C1(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5063 to 1.5083 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only