N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
Thermo Scientific Chemicals

N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%

CAS: 140-07-8 | C10H24N2O4 | 236.31 g/mol
数量:
100 g
500 g
1 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 420481000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS140-07-8
IUPAC Name2-({2-[bis(2-hydroxyethyl)amino]ethyl}(2-hydroxyethyl)amino)ethan-1-ol
Molecular FormulaC10H24N2O4
InChI KeyBYACHAOCSIPLCM-UHFFFAOYSA-N
SMILESOCCN(CCO)CCN(CCO)CCO
さらに表示
Appearance (Color)Clear colorless to amber
Titration with HClO4>=98.5 %
Refractive index1.4990 to 1.5070 (20°C, 589 nm)
Appearance (Form)Viscous liquid
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only