N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
Thermo Scientific Chemicals

N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%

CAS: 637-01-4 | C10H16N2·2HCl | 237.17 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 420601000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS637-01-4
IUPAC Namedihydrogen N1,N1,N4,N4-tetramethylbenzene-1,4-diamine dichloride
Molecular FormulaC10H18Cl2N2
InChI KeyFBHKTSXMTASXFJ-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].CN(C)C1=CC=C(C=C1)N(C)C
さらに表示
Appearance (Color)White to beige to grey to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO498.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only