Thymolphthalein, ACS reagent
Thymolphthalein, ACS reagent
Thymolphthalein, ACS reagent
Thermo Scientific Chemicals

Thymolphthalein, ACS reagent

CAS: 125-20-2 | C28H30O4 | 430.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 420950100
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.90
Save 6.25
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.90
Save 6.25
Each
Chemical Identifiers
CAS125-20-2
IUPAC Name3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC28H30O4
InChI KeyLDKDGDIWEUUXSH-UHFFFAOYSA-N
SMILESCC(C)C1=CC(=C(C)C=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Visual transition intervalfrom pH 8.8 (colorless) to pH 10.5 (blue)
Appearance (Form)Crystalline powder
Clarity of solutionpasses test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only