D-delta-Tocopherol, 93%
D-delta-Tocopherol, 93%
D-delta-Tocopherol, 93%
Thermo Scientific Chemicals

D-delta-Tocopherol, 93%

CAS: 119-13-1 | C27H46O2 | 402.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Catalog number 421051000
Price (USD)
255.65
Special offer
Online exclusive
Ends: 28-Dec-2026
300.00
Save 44.35
Each
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
255.65
Special offer
Online exclusive
Ends: 28-Dec-2026
300.00
Save 44.35
Each
Chemical Identifiers
CAS119-13-1
IUPAC Name(2R)-2,8-dimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Molecular FormulaC27H46O2
InChI KeyGZIFEOYASATJEH-VHFRWLAGSA-N
SMILESCC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=CC(C)=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
GC>=91 %
Infrared spectrumConforms
Appearance (Color)Clear yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only